Cynomolgus monkey Recombinant B7-H3/CD276 Protein (from HEK293 cells)
Catalog # 77130-204
Supplier: Prosci
CAS Number:
Specifications
- Pack type:Vial
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Cynomolgus monkey
- Size:0.1 mg
- Environmentally Preferable:
- Storage conditions:Lyophilized protein should be stored at −20 °C or lower for long term storage. Upon reconstitution, working aliquots should be stored at −20 or −70 °C. Avoid repeated freeze-thaw cycles.
- Gene ID:102131295
- Protein synonyms:B7 homolog 3|B7-H3|CD276
- Protein/peptide name:CD276
- Purity:>95% as determined by SDS-PAGE
- Molecular weight:49.0 kDa
- Sequence:Leu 29 - Glu 465
- Formulation:Lyophilized. PBS, pH 7.4.
- Shipping temperature:RT
- Tested applications:ELISA,WB
- Cat. no.:77130-204