SARS-CoV-2 (COVID-19) Recombinant S Protein RBD (from HEK293 cells)
Catalog #: 77132-114
Supplier: Prosci
New Product
CAS Number:
Specifications
- Gene ID:43740568
- Packaging:Vial
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:SARS-CoV-2
- Conjugation:Unconjugated
- Size:0.2 mg
- Storage conditions:Lyophilized protein should be stored at −20 °C or lower for long term storage. Upon reconstitution, working aliquots should be stored at −20 or −70 °C. Avoid repeated freeze-thaw cycles.
- Protein synonyms:SpikeS glycoprotein RBDSpike glycoprotein Receptor-binding domainSpike protein RBDS protein RBDCOVID-19
- Protein/peptide name:Spike
- Purity:>95% as determined by SDS-PAGE
- Molecular weight:27.0 kDa
- Formulation:Liquid. 10 mM PB, pH 7.4.
- Shipping temperature:Dry ice
- Tested applications:ELISA,WB
Specifications
About this item
Customers Who Bought This Also Bought