Cynomolgus monkey Recombinant TREM2 Protein (from HEK293 cells)
Catalog # 77121-956
Supplier: Prosci
CAS Number:
Specifications
- Pack type:Vial
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Cynomolgus monkey
- Size:0.1 mg
- Environmentally Preferable:
- Storage conditions:Lyophilized protein should be stored at −20 °C or lower for long term storage. Upon reconstitution, working aliquots should be stored at −20 or −70 °C. Avoid repeated freeze-thaw cycles.
- Gene ID:102133279
- Protein synonyms:Triggering receptor expressed on myeloid cells 2/TREM2/TREM-2
- Protein/peptide name:TREM2
- Purity:>90% as determined by SDS-PAGE
- Molecular weight:19.3 kDa
- Sequence:His 19 - Ser 174
- Formulation:Lyophilized. PBS, pH 7.4.
- Shipping temperature:RT
- Tested applications:ELISA,WB
- Cat. no.:77121-956