Human Recombinant ACE2 Protein (from Mammalian Cells)
77135-052
: Prosci
:
- Pack type:Vial
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:Mammalian cells
- Species:Human
- Size:0.1 mg
- Storage conditions:Use a manual defrost freezer and avoid repeated freeze thaw cycles. Store at 2...8 °C for one week. Store at −20...−80 °C for twelve months from the date of receipt.
- Protein synonyms:Angiotensin-converting enzyme homolog|Metalloprotease MPROT15|ACE-related carboxypeptidase
- Protein/peptide name:ACE2
- Purity:>90% as determined by SDS-PAGE
- Molecular weight:85.2 kDa
- Sequence:Met1-Ser740
- Formulation:Lyophilized. Supplied as Lyophilized from TBS, pH 8.0. Please reconstitute the protein with ddH₂O to the concentration with 1 mg/ml.
- Shipping temperature:RT
- Cat. no.:77135-052