SARS-CoV-2 (COVID-19) Recombinant Spike S1 Protein (from HEK293 cells)
77134-684
: Prosci
:
- Pack type:Vial
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:SARS-CoV-2
- Size:0.1 mg
- Storage conditions:The protein is stable in a liquid state at −70 °C for 12 months. Avoid repeated freeze-thaw cycles.
- Protein synonyms:Spike RBD Protein|ncov RBD Protein|S1 Protein|RBD Protein|ncov s2 Protein|S2 Protein|cov spike Protein|ncov s1 Protein|SARS-CoV-2 S|coronavirus spike Protein|novel coronavirus spike Protein
- Protein/peptide name:Spike S1
- Purity:<95%
- Molecular weight:76.2 kDa
- Sequence:Val16-Arg685
- Concentration:1 mg/ml
- Formulation:Liquid, PBS.
- Shipping temperature:Dry Ice
- Tested applications:ELISA,WB
- Cat. no.:77134-684