Anti-ADRB2 Recombinant Antibody [Clone: 1B1.3a]
Catalog # 76629-302
Supplier: Prosci
Specifications
- Antibody type:Primary
- Antigen name:adrenoceptor beta 2
- Antigen symbol:ADRB2
- Clonality:Recombinant
- Clone:1B1.3a
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Isotype:IgG1 kappa
- Reactivity:Mouse
- Size:0.2 mg
- Cross adsorption:No
- Form:Liquid
- Gene ID:16154
- Antigen synonyms:Interleukin-10 receptor subunit|CD210|Interleukin-10 receptor subunit alpha|CDw210a|IBI-3A|IL-10R subunit alpha|IL-10 receptor subunit alpha| Il10ra Il10r|1B1-3A|IL-10R subunit 1|Interleukin-10 receptor subunit 1|IL-10RA|IL-10R1
- Modification:Unmodified
- Storage buffer:PBS with 0.02% Proclin 300
- Storage temperature:Store at 4 °C for up to 3 months. For longer storage, aliquot and store at −20 °C.
- Shipping temperature:Shipped on blue ice
- Immunogen:This antibody was raised by immunising rats with the purified recombinant extracellular region of mouse Interleukin 10 Receptor.
- Tested applications:Blocking
- Cat. no.:76629-302
- Packaging:Vial
Specifications
About this item
Type: Primary
Antigen: adrenoceptor beta 2
Clonality: Recombinant
Clone: 1B1.3a
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG1 kappa
Reactivity: Mouse