Anti-RAP Recombinant Antibody [Clone: 7F1]
Catalog # 76630-600
Supplier: Prosci
Specifications
- Antibody type:Primary
- Clonality:Recombinant
- Clone:7F1
- Conjugation:Unconjugated
- ELISA:Yes
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG kappa
- Reactivity:Human
- Western blot:Yes
- Size:0.2 mg
- Cross adsorption:No
- Form:Liquid
- Antigen synonyms:low density lipoprotein receptor-associated protein|Receptor-associated protein|39-kDa receptor-associated protein
- Modification:Unmodified
- Storage buffer:PBS with 0.02% Proclin 300
- Storage temperature:Store at 4 °C for up to 3 months. For longer storage, aliquot and store at −20 °C.
- Shipping temperature:Shipped on blue ice
- Immunogen:This antibody was raised by immunising mice with human placental RAP.
- Cat. no.:76630-600
- Packaging:Vial
Specifications
About this item
Type: Primary
Antigen: RAP
Clonality: Recombinant
Clone: 7F1
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG kappa
Reactivity: Human