Anti-ITGAM Recombinant Antibody [Clone: 5C6 (recombinant version)]
76629-926
: Prosci
- Antibody type:Primary
- Antigen name:integrin subunit alpha M
- Antigen symbol:ITGAM
- Clonality:Recombinant
- Clone:5C6 (recombinant version)
- Conjugation:Unconjugated
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG2b kappa
- Reactivity:Human,Mouse
- Size:0.1 mg
- Cross adsorption:No
- Form:Liquid
- Gene ID:3684
- Antigen synonyms:Neutrophil adherence receptor|Cell surface glycoprotein MAC-1 subunit alpha|CD11 antigen-like family member B|CR-3 alpha chain|Leukocyte adhesion receptor MO1|integrin alpha M
- Modification:Unmodified
- Storage buffer:PBS with 0.02% Proclin 300
- Storage temperature:Store at 4 °C for up to 3 months. For longer storage, aliquot and store at −20 °C.
- Shipping temperature:Shipped on blue ice
- Immunogen:Fusion of spleen cells from AO rats immunised with Thioglycollate-elicted muine peritoneal macrophages (TPM) and the Y3 rat myeloma line.
- Cat. no.:76629-926
- Packaging:Vial
Type: Primary
Antigen: integrin subunit alpha M
Clonality: Recombinant
Clone: 5C6 (recombinant version)
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG2b kappa
Reactivity: Mouse;Human