Anti-Cd3e Recombinant Antibody [Clone: 145-2C11]
76629-628
: Prosci
- Antibody type:Primary
- Antigen name:CD3 antigen, epsilon polypeptide
- Antigen symbol:Cd3e
- Clonality:Recombinant
- Clone:145-2C11
- Conjugation:Unconjugated
- Flow cytometry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG kappa
- Reactivity:Mouse
- Western blot:Yes
- Size:0.2 mg
- Cross adsorption:No
- Form:Liquid
- Gene ID:12501
- Antigen synonyms:CD3e|T-cell surface antigen T3/Leu-4 epsilon chain CD_antigen|T-cell surface glycoprotein CD3 epsilon chain
- Modification:Unmodified
- Storage buffer:PBS with 0.02% Proclin 300
- Storage temperature:Store at 4 °C for up to 3 months. For longer storage, aliquot and store at −20 °C.
- Shipping temperature:Shipped on blue ice
- Immunogen:H-2Kb-specific murine cytotoxic T-lymphocyte (CTL) clone.
- Tested applications:Activation
- Cat. no.:76629-628
- Packaging:Vial
Type: Primary
Antigen: CD3 antigen, epsilon polypeptide
Clonality: Recombinant
Clone: 145-2C11
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG kappa
Reactivity: Mouse