Anti-LTA Recombinant Antibody [Clone: 3F12.2D3]
Catalog # 76630-002
Supplier: Prosci
Specifications
- Antibody type:Primary
- Antigen name:lymphotoxin alpha
- Antigen symbol:LTA
- Clonality:Recombinant
- Clone:3F12.2D3
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Isotype:IgG2b kappa
- Reactivity:Human
- Size:0.2 mg
- Cross adsorption:No
- Neutralisation:Yes
- Form:Liquid
- Gene ID:4049
- Antigen synonyms:Lymphotoxin-alpha|TNF-beta|TNF β|Lymphotoxin alpha|Tumor necrosis factor β|Tumor necrosis factor-beta|TNF beta|TNF-β|Tumor necrosis factor beta|LT|LTα3 chimeric 3F12|LTα|tumor necrosis factor-β
- Modification:Unmodified
- Storage buffer:PBS with 0.02% Proclin 300
- Storage temperature:Store at 4 °C for up to 3 months. For longer storage, aliquot and store at −20 °C.
- Shipping temperature:Shipped on blue ice
- Immunogen:3F12 was prepared by hyperimmunizing BALB/c mice with purified recombinant human LTα expressed in E. coli - the hybridoma clone was selected for its ability to bind LTα3 by ELISA. The CDRs of the antibody were grafted to the framework regions of the mice mAb 2G7 to create the chimeric version of the antibody.
- Tested applications:Surface Plasma Resonance
- Cat. no.:76630-002
- Packaging:Vial
Specifications
About this item
Type: Primary
Antigen: lymphotoxin alpha
Clonality: Recombinant
Clone: 3F12.2D3
Conjugation: Unconjugated
Host: Recombinant
Isotype: IgG2b kappa
Reactivity: Human