Eisco Round Bottom Flask Stands
Supplier: Wards
Certificates
About this item
Cup-shaped flask stand capable of holding up to 10 L round bottom flasks upright.
- Sturdy polypropylene plastic construction will stand up to the wear and tear of the lab
- Cup-shaped, multi-level flask placement decks accommodate flasks of various shapes and sizes
- Diameters of multi-level decks range from 1 to 6.25"
- Can hold a single round bottom flask up to 10 L capacity
- Stackable design for convenient storage
- Can be autoclaved
The benchtop flask holder is made of sturdy polypropylene plastic and has multi-leveled flask placement decks ranging from a diameter of 1 to 6.25". This cup-shaped, multi-level design accommodates flasks of various shapes and sizes. Overall size: 6.5Ø×2"H.
Frequently Bought Together