Specifications
- Clothing Size:One size
- Color:Blue
- Cat. no.:77148-078
Specifications
About this item
Disposable shoe covers can be worn where there is a likelihood of blood, bodily fluids, secretions, or excretions splashing or spilling on the shoes. RightCare shoe covers are fluid-resistant and chemical-resistant so the physical properties of the covers are not affected by splashes of solvents.
- CPE material contains dirt particles while also repelling moisture and oil
- Disposable shoe covers should not be reused
- Heavily textured soles allow for more grip to reduce the risk of slips, falls, and injuries
- Seamless bottoms
- 16" length, 6" height
- Nonsterile
RightCare shoe covers are designed to contain moisture and dirt, protecting both shoes and the environment. Constructed from a durable mixture of 65% low-density polyethylene (LDPE) and 35% light low-density polyethylene (LLDPE), the material is also known as polylatex or CPE. Heavily textured soles allow for more grip to reduce the risk of slips, falls, and injuries.
Polylatex has excellent resistance to corrosion making these covers suitable for various environments such as medical facilities, laboratories, pharmacies, construction sites, automotive, schools, businesses, or even at home. These are also commonly used for real estate tours of homes.