Inverted Microscope with FOV20 and S40 Basic Stand Outfit, DMi1
Catalog #: CA76538-908
Supplier: Leica Microsystems
Specifications
- Description:Inverted tissue culture microscope with 20 mm eyepieces, 40 mm WD condenser
- Cat. no.:CA76538-908
Specifications
About this item
Microscope for basic cell culture applications.
- Two eyepieces (10×/20 eyepiece with eyeguard and 10×/20 focusing eyepiece with eyeguard)
- S40 condenser
- 4 position slider for S40 condenser (BF, Ph0, Ph1, Ph2)
- 3 objectives (HI PLAN 4×/0.10 PH0, HI PLAN I 10×/0.22 PH1, HI PLAN I 20×/0.30 PH1)
- Dust cover
Leica DMi1 FOV20 basic stand (FOV20, LED illumination, exchangeable eyepieces, fixed stage, power cable).
Customers Who Bought This Also Bought