About this item
Foxx Life Sciences new EZBio®pure Titanium Round (TR) media bottles have enhanced the media bottle design. The improved design features an angled shoulder to ensure smooth and complete media pouring.
- Media bottles available in 125, 250, 500 and 1000 ml sizes
- Made entirely of 100% USP Class VI components
- Compatible with leakproof GL45 VersaCaps®, for flexibility of interchangeable connectors
- Bottles can withstand low temperatures down to −70 ºC
- Manufactured in a Class 7 cleanroom
The new titanium round media bottles are perfect for storing and sampling active pharmaceutical ingredients, bulk intermediates, and preparing and storing buffers for culture. These bottles can also be used for long-term storage of pH sensitive liquids. Molded finger grips improve the ability to grasp a bottle, even with gloved hands. The lower height enhances stability, and bottles also feature molded graduation marks that are easy to read. EZBio®pure TR Bottles are made of Polyethylene Terephthalate Glycol (PETG) resin. The crystal clear material uses USP Class VI resins.