Orders LinkContactUsLinkComponent
Quick-DNA/RNA™ Miniprep Plus Kits
Quick-DNA/RNA™ Miniprep Plus Kits
Supplier:   Zymo Research
Quick-DNA/RNA™ Miniprep Plus Kits
Supplier:  Zymo Research
Certificates
Quick-DNA/RNA™ Miniprep Plus Kits
2 Options Available Below
Product Details & Documents
The Quick-DNA/RNA MiniPrep Plus kit combines Quick-DNA/RNA technology with the addition of DNA/RNA Shield, a preservation and lysis technology, and Proteinase K to enable easy, reliable, and rapid isolation from any biological sample including cells, solid tissue, and whole blood. The procedure uses Zymo-Spin column technology that results in high-quality genomic DNA and total RNA that is ready for any downstream application including reverse transcription, microarray, sequencing, etc.
Specifications for Product Family
Specifications
Specifications
  • Catalog No:
  • 77001-854
  • 76358-832
  • Binding capacity:
  • 100 µg
  • 100 µg
  • Yield:
  • 100 µg DNA and RNA (binding capacity), ≥50 µl (elution volume)
  • 100 µg DNA and RNA (binding capacity), ≥50 µl (elution volume)
Specifications
Product Family Options
Product Information
  • Details
    Size
    Availability
    Price
  • 77001-854
    Quick-DNA/RNA™ Miniprep Plus
    Quick-DNA/RNA™ Miniprep Plus
    for Cells, any tissues, blood, biological fluids
    50 Preps
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - 765,76 $
    765,76 $
    Log in for availability
    Kit of 1
    Kit of 1 - 765,76 $
    765,76 $
    Each
       765,76 $
    Minimum order quantity is 1
    Kit of 1
       765,76 $
    Minimum order quantity is 1
    Supplier Number: D7003
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    More
    Specifications
    Cat. No.
    77001-854
    Binding capacity
    100 µg
    Yield
    100 µg DNA and RNA (binding capacity), ≥50 µl (elution volume)
  • 76358-832
    Quick-DNA/RNA™ Miniprep Plus
    Quick-DNA/RNA™ Miniprep Plus
    for Cells, any tissues, blood, biological fluids
    10 Preps
    Log in for availability
    Log in for availability
    Each
    Each - 186,65 $
    186,65 $
    Each
       186,65 $
    Supplier Number: D7003T
    Specifications:
    More
    Specifications
    Cat. No.
    76358-832
    Binding capacity
    100 µg
    Yield
    100 µg DNA and RNA (binding capacity), ≥50 µl (elution volume)