PIP® Glove Inflator Kit
76944-990
: PIP
- Description:Glove inflator kit
- Sterility:Non sterile
- Cat. no.:76944-990
Glove inflator for inspecting rubber gloves.
- Easy-to-use design with a non-slip base
- No straps or bands required
- Use for quick and thorough inspection of insulated rubber gloves
- Will fit all Novax insulated gloves
The easy-to-use design features a non-slip base and doesn't require straps or bands for fast and thorough visual inspections, which are required prior to each use of the insulating gloves.
: In compliance with OSHA 1910.137 requiring daily inspection of insulating equipment.