CleanTeam® Nylon/Copper Fiber ESD Gloves
Supplier: PIP
New Product
Certificates
About this item
Seamless knit nylon/copper fiber electrostatic dissipative (ESD) gloves, with a polyurethane-coated smooth grip available on the fingertips, or both the palm and full fingers.
- Electrostatic dissipative (ESD) copper fiber yarns blended with low-lint nylon
- Reduced static build-up on the glove surface for improved performance in electronics assembly
- Seamless knit gloves provide great dexterity
- Reusable and launderable
Ideally suited for use in circuit board manufacturing, munitions, electronics assembly and aerospace applications. The smooth polyurethane coating provides a secure grip in dry and slightly wet/oily conditions, and offers high dexterity and tactile sensitivity.
Packaging: Gloves are packaged 25 dozen per case.
Specifications
- Catalog No:
- 76944-966
- 76944-964
- 76944-962
- 76944-960
- Ambidextrous/Hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Latex/Latex-free:
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Powdered/Powder-free:
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Gauge:
- 13
- 13
- 13
- 13
- Cuff style:
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Sterility:
- Non sterile
- Non sterile
- Non sterile
- Non sterile