CleanTeam® Nylon/Carbon Fiber ESD Gloves
Supplier: PIP
New Product
Certificates
About this item
Seamless knit nylon/carbon fiber electrostatic dissipative (ESD) gloves, available with or without raised PVC dot grip enhancement.
- Electrostatic dissipative (ESD) carbon fiber yarns blended with low-lint nylon
- Reduced static build-up on the glove surface for improved performance in electronics assembly
- Seamless knit gloves provide great dexterity
- Reusable and launderable
Ideally suited for use in circuit board manufacturing, munitions, electronics assembly, and aerospace applications. Gloves featuring raised PVC dots provide enhanced grip and extra cut and abrasion resistance.
Packaging: Gloves are packaged 25 dozen per case.
Specifications
- Catalog No:
- 76944-956
- 76944-958
- 76944-952
- 76944-954
- Ambidextrous/Hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Latex/Latex-free:
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Powdered/Powder-free:
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Gauge:
- 13
- 13
- 13
- 13
- Cuff style:
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Sterility:
- Non sterile
- Non sterile
- Non sterile
- Non sterile