MaxiFlex® Elite™ Anti-Static Nylon Gloves with Nitrile Coating
: PIP
Anti-static seamless knit nylon glove with nitrile coated MicroFoam grip on palm and fingers.
- Ultra light-weight nylon liner creates smooth and rounded fingertips maximizing sensitivity, dexterity and tactility
- 30% thinner than traditional foam nitrile gloves
- Touchscreen compatible; operate a touch screen phone or device without removing gloves
- Patented nitrile MicroFoam coating offers 360° breathability
- Minimal penetration (strike through), leaving only the soft liner resting against the skin
- Silicone-free and latex-free
Weighing in at a significant 36% less and 30% thinner than traditional foam nitrile gloves MaxiFlex® Elite™ delivers a user experience that sets new standards. Comfort is further enhanced through their increased breathability, which is up to 40% more than similar gloves.
: Complies with EN 16350-2014 for electrostatic protective gloves. Also tested to EN 61340-2-3:2017 for achieving anti-static properties with vertical resistance.
: Gloves are packaged 12 dozen per case.
- Catalog No:
- 76944-812
- 76944-818
- 76944-816
- 76944-814
- 76944-820
- 76944-810
- Ambidextrous/Hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Latex/Latex-free:
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Powdered/Powder-free:
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Finish:
- MicroFoam grip
- MicroFoam grip
- MicroFoam grip
- MicroFoam grip
- MicroFoam grip
- MicroFoam grip
- Gauge:
- 18
- 18
- 18
- 18
- 18
- 18
- Cuff style:
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Sterility:
- Non sterile
- Non sterile
- Non sterile
- Non sterile
- Non sterile
- Non sterile
Frequently Bought Together