Order Entry
Canada
Orders LinkContactUsLinkComponent
Hardshell Plastic Cuvette Cases, Type P

Hardshell Plastic Cuvette Cases, Type P

Supplier:   FIREFLYSCI, INC.
Hardshell Plastic Cuvette Cases, Type P
Supplier:  FIREFLYSCI, INC.
Certificates
Hardshell Plastic Cuvette Cases, Type P
Hardshell Plastic Cuvette Cases, Type P

1 Options Available Below

Product Details & Documents

Cuvette cases hold 4 cells up to 10 mm L/P each, or two 20 mm L/P cells, or one 50 mm L/P cell.

  • Black opaque finish
Product Family Options

Product Information

  • Material
    Availability
    Price
  • 76016-134
    Cuvette case for 45 mm cells
    Cuvette case for 45 mm cells
    Polyfoam
    Discontinued
    Supplier Number: -
  • 76016-136
    Cuvette case for 49 - 55 mm cells
    Cuvette case for 49 - 55 mm cells
    Polyfoam
    Discontinued
    Supplier Number: -
  • 76016-652
    Cuvette case for 45 - 55 mm cells
    Cuvette case for 45 - 55 mm cells
    Plastic
    Log in for availability
    Log in for availability
    Each
    Each - $75.09
    $75.09
    Each
       $75.09

    Supplier Number: CASEP4
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.