Sub-Micro Black Fluorometer Cuvette (Capacity: 10 - 160 µl), Type 701MFL
Supplier: FIREFLYSCI, INC.
Certificates
About this item
FireflySci's 10 mm sub-micro fluorometer cuvette with capacities ranging from 10 to 160 µl.
- Fire-fused to ensure that cuvettes are robust and durable
- Available in UV quartz
- Capacities of 10, 50, 100, 160 µl
- Three windows optically polished for 90º emission/excitation applications
- Includes PTFE threaded stopper
Great for DNA, RNA, and Protein micro samples.
Customers Who Bought This Also Bought