Semi-Micro Fluorescence Cuvette with PTFE Cover (Lightpath: 10 mm), Type 9FL
Supplier: FIREFLYSCI, INC.
Certificates
About this item
Standard semi-micro fluorometer cuvette with PTFE cover.
- Fire-fused to ensure that cuvettes are robust and durable
- Available in several different materials: Optical glass, UV quartz, IR quartz
- Available in 10 mm lightpath
- Four windows and base optically polished
- Includes PTFE cover
Customers Who Bought This Also Bought