Semi-Micro Cuvette with PTFE Cover (Lightpaths 5 - 100 mm), Type 9
Supplier: FIREFLYSCI, INC.
Certificates
About this item
Standard semi-micro spectrophotometer cuvette with PTFE cover.
- Available in several different materials: Optical glass, UV quartz, IR quartz
- Excellent transmission values
- Lightpaths from 5 to 100 mm are available
- Fire-fused to ensure that cuvettes are robust and durable
Cuvettes provide highly accurate results, meaning users only need to test a sample once. Excellent transmission values.
Frequently Bought Together