Coverslips
Mopec coverslips are made with borosilicate glass.
- Borosilicate glass, #1 thickness, 12 mm in diameter
- Will not stick or auto-fluoresce
- Clean, uniform surface
- Ideal for automated coverslipping
Available in 1 or 2 ounce volumes.
: The coverslips are packaged 10 boxes per case, with 100 or 200 pieces per box.
Mopec coverslips are made with borosilicate glass.
- Borosilicate glass, #1 thickness, 12 mm in diameter
- Will not stick or auto-fluoresce
- Clean, uniform surface
- Ideal for automated coverslipping
Available in 1 or 2 ounce volumes.
: The coverslips are packaged 10 boxes per case, with 100 or 200 pieces per box.
Product Family Options
- L×WVolume
- 24×50 mm1 oz.-: SL024-50-10
- : SL024-50-20
- : SL024-55-10
- : SL024-55-20
- : SL024-60-10
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)