M6 Threaded Connectors for Tablet Dissolution Cells, A19
76016-678
- Description:M6 threaded connectors
- Cat. no.:76016-678
A pair of M6 threaded connectors for use in Type 601, 602, and 606 tablet dissolution cells. The connector holds the PTFE tube firmly in place to seal it tightly against the quartz cell.
- The dimensions of the supplied tubing is OD: 1.6 mm, ID: 0.8 mm. The length is 400 mm