Order Entry
Canada
ContactUsLinkComponent
Cuvette Mount (For Type 507 and 607 Cuvettes), A24
Cuvette Mount (For Type 507 and 607 Cuvettes), A24
Catalog # 76016-662
Supplier:  FIREFLYSCI, INC.
undefined
Cuvette Mount (For Type 507 and 607 Cuvettes), A24
Catalog # 76016-662
Supplier:  FIREFLYSCI, INC.
Supplier Number:  A24

Specifications

  • Description:
    Cuvette mount for Type 507 and 607 cuvettes
  • Cat. no.:
    76016-662

Specifications

About this item

Cell mount for Type 507 and Type 607 micro cells.

  • Anodized aluminum construction
  • External dimensions: 12.5L×12.5W×43H mm

Mount enables Type 507 and Type 607 cuvettes to fit in a standard 10 mm cuvette holder.