Orders LinkContactUsLinkComponent
Cuvette Rack, Type P66
Cuvette Rack, Type P66
Polyethylene cuvette rack
Catalog # 76016-654
Supplier:   FIREFLYSCI, INC.
Supplier #:  P66
Cuvette Rack, Type P66
Catalog # 76016-654
Supplier:  FIREFLYSCI, INC.
Cuvette Rack, Type P66
Cuvette Rack, Type P66
Polyethylene cuvette rack
Price
$325.35
Log in for availability
Each
Product Details & Documents
Cell holder rack for 12 standard size cuvettes.
  • 12-hole cell rack
  • Designed to hold twelve (12) 10 mm pathlength cells
Rack is manufactured from polyethylene to ensure that windows cannot be scratched or damaged when being placed in the rack.
Specifications
  • Description:
    Polyethylene cuvette rack