Triangular Fluorescence Cell with PTFE Stopper (Lightpath: 10 mm), Type 82
Marketsource
Supplier: FIREFLYSCI, INC.
Triangular Fluorescence Cell with PTFE Stopper (Lightpath: 10 mm), Type 82
Supplier: FIREFLYSCI, INC.
Marketsource
Certificates
Jump to:
About this item
Triangular fluorescence cell with PTFE stopper.
- Fire fused to ensure the best bonding
- Different materials available: UV Quartz and IR Quartz
- Available in 10 mm lightpath
- Angles: 45º×45º×90º
- Comes with PTFE stopper.
Three windows and the base are optically polished.
2 Options Available Below
Product Details & Documents
Triangular fluorescence cell with PTFE stopper.
- Fire fused to ensure the best bonding
- Different materials available: UV Quartz and IR Quartz
- Available in 10 mm lightpath
- Angles: 45º×45º×90º
- Comes with PTFE stopper.
Three windows and the base are optically polished.
Product Family Options
Product Information
- CapacityInternal dimensionsExternal dimensionsLightpathMaterialAvailabilityPrice
- 1.750 mlL×W×H 10×10×45 mmL×W×H 12.5×12.5×49 mm10 mmUV Quartz (190 - 2500 nm)Log in for availabilityEach$1,458.84Supplier Number: 82UV10
- 1.750 mlL×W×H 10×10×45 mmL×W×H 12.5×12.5×49 mm10 mmIR Quartz (220 - 3500 nm)Log in for availabilityEach$1,163.26Supplier Number: 82IR10Specifications:MarketsourceMarketsource
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



