Semi-Micro Raised-Bottom Black Cuvette with PTFE Cover (Lightpaths: 10×1 to 10×5 mm), Type 9BM
Supplier: FIREFLYSCI, INC.
Certificates
About this item
Semi-micro spectrophotometer cuvette with PTFE cover, made with black quartz.
- Fire-fused to ensure that cuvettes are robust and durable
- Available in UV Quartz and IR Quartz
- Lightpaths of 10 mm available with various inner widths (10×1 to 10×5 mm)
- Base thickness: 9 mm
- Two windows optically polished
- Walls are made from black quartz to block out stray light
- Includes PTFE cover
Floor of the cuvette is raised 9 mm.
Customers Who Bought This Also Bought