Orders LinkContactUsLinkComponent
Sampling Scoops, PS
Sampling Scoops, PS
Sampling Scoops, PS
Supplier:  BURKLE INC
Certificates
Sampling Scoops, PS
Sampling Scoops, PS
4 Options Available Below
Product Details & Documents
Scoops are ideal for the sampling of powder, granules and pastes.
  • Individually packed, gamma radiation sterilized
Special handle construction and the slightly angled edge stands horizontal and can be laid down without the material trickling out.
Certifications: Production, assembling and packaging according to cleanroom Class 7 (10,000). Complies with food and FDA guidelines.
Product Family Options
Product Information
  • Capacity
    Length
    Color
    Availability
    Price
  • Sampling Scoops
    25 ml (0.85 oz.)
    141 mm (5.55")
    White
    Log in for availability
    Log in for availability
    Pack of 100
    Pack of 100 - $393.35
    $393.35
    Pack of 100
       $393.35
    Supplier Number: 5378-9906
  • Sampling Scoops
    100 ml (3.38 oz.)
    205 mm (8.07")
    White
    Log in for availability
    Log in for availability
    Pack of 100
    Pack of 100 - $521.45
    $521.45
    Pack of 100
       $521.45
    Supplier Number: 5378-9907
  • Sampling Scoops
    250 ml (8.45 oz.)
    232 mm (9.13")
    White
    Log in for availability
    Log in for availability
    Pack of 100
    Pack of 100 - $724.05
    $724.05
    Pack of 100
       $724.05
    Supplier Number: 5378-9908
  • Sampling Scoops
    500 ml (16.91 oz.)
    280 mm (11.02")
    White
    Log in for availability
    Log in for availability
    Pack of 100
    Pack of 100 - $1,298.25
    $1,298.25
    Pack of 100
       $1,298.25
    Supplier Number: 5378-9910