Order Entry
Canada
ContactUsLinkComponent
Universal Laboratory Hydrometer
Universal Laboratory Hydrometer
Catalog # 470344-922
Supplier:  Wards
undefined
Universal Laboratory Hydrometer
Catalog # 470344-922
Supplier:  Wards
Supplier Number:  PH0128

Specifications

  • Description:
    Universal laboratory hydrometer
  • Length:
    15.25"
  • Cat. no.:
    470344-922
  • Stem O.D.:
    0.58"
  • Graduations:
    0.01
  • Range:
    0.700 - 2.000

Specifications

About this item

Measures specific gravity range of 0.700 to 2.000 with 0.01 graduations and dual scale for light and heavy liquids.

  • Universal laboratory hydrometer in hard plastic case measure specific gravity range 0.700 to 2.000
  • Dual scale hydrometer designed for lab use
  • Can be used for beer, wine, kombucha, mead, and other specific gravity activities
  • Designed to be used in the laboratory, but can be used in the home or garage as well

Lab quality hydrometer with dual scale.