ContactUsLinkComponent
Control Slides, Tissue Microarray, Normal Human Tissue, 8 Tissue Types
Control Slides, Tissue Microarray, Normal Human Tissue, 8 Tissue Types
Catalog # 76948-258
Supplier:  Mopec
undefined
Control Slides, Tissue Microarray, Normal Human Tissue, 8 Tissue Types
Catalog # 76948-258
Supplier:  Mopec
Supplier Number:  SC982-25
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    Mopec control slides, tissue microarray, normal human tissue, 8 tissue types
  • Cat. no.:
    76948-258
Specifications
About this item
All control slides are on Mopec IHC enhanced hydrophilic adhesive slides using Formalin Fixed Paraffin Embedded (FFPE) human tissue.
  • Product specific reactivity is guaranteed for one year from date of receipt
  • Revalidate after one year to verify continued reactivity
  • All controls are cut in serial sections at 4 µ
  • Standard tissue placement is mid-high on slide to facilitate placement of patient and control tissue on the same slide
  • All slides are air dried
Product specific reactivity is guaranteed for one year from date of receipt when stored properly at 60 to 80 °F in a humidity controlled and light-deprived environment.
Packaging: Tissue type: chosen from liver, lung, kidney, colon, tonsil, skin, prostate, breast, lymph node, uterus, testis, placenta, thyroid, stomach, and pancreas.