Orders LinkContactUsLinkComponent
Eisco Fixed Volume Micropipettes
Eisco Fixed Volume Micropipettes
Supplier:   Wards
Eisco Fixed Volume Micropipettes
Supplier:  Wards
Certificates
Eisco Fixed Volume Micropipettes
Eisco Fixed Volume Micropipettes
14 Options Available Below
Product Details & Documents
Factory-calibrated, color-coded Fixed Volume Micropipettes with tip ejector, PVDF cone, and body grip conform to DIN 12650 and EN ISO 8655 standards.
  • Fixed volume micropipettes
  • Features a soft and lightweight operation with an ergonomic, soft touch tip ejector designed to minimize hand fatigue, reduce thumb stress and to increase comfort with prolonged use
  • Fully autoclavable with a durable, autoclavable PVDF tip cone and a slip-resistant thermoplastic elastomer (TPE) body grip to minimize the transfer of body heat to the micropipette resulting in high accuracy even with continuous use
  • Entire series of micropipettes features color coded plungers for easy identification
  • Calibration conforms to DIN 12650 and EN ISO 8655 standard ensuring high accuracy and precision
  • Calibration report enclosed with every micropipette
High performance fixed volume micropipettes have a simple design which allows for easy use. Features soft and lightweight operation with an ergonomic, soft touch tip ejector designed to minimize hand fatigue, reduce thumb stress and to increase comfort with prolonged use.
Fully autoclavable with a durable, autoclavable PVDF tip cone and a slip-resistant thermoplastic elastomer (TPE) body grip to minimize the transfer of body heat to the micropipette resulting in high accuracy even with continuous use.
The entire series of Eisco Fixed Volume Micropipettes features color coded plungers for easy identification. Calibration conforms to DIN 12650 and EN ISO 8655 standard ensuring high accuracy and precision. Calibration report enclosed with every micropipette.
`
Product Family Options
Product Information
  • Volume
    Color code
    Availability
    Price
  • 470343-746
    Micropipette
    Micropipette
    2.5 µl
    Red
    Log in for availability
    Log in for availability
    Each
    Each - $182.25
    $182.25
    Each
       $182.25
    Supplier Number: FVM01
  • 470346-092
    Micropipette
    Micropipette
    5 µl
    Red
    Log in for availability
    Log in for availability
    Each
    Each - $185.95
    $185.95
    Each
       $185.95
    Supplier Number: FVM02
  • 470343-936
    Micropipette
    Micropipette
    10 µl
    Black
    Log in for availability
    Log in for availability
    Each
    Each - $167.05
    $167.05
    Each
       $167.05
    Supplier Number: FVM03
  • 470344-536
    Micropipette
    Micropipette
    20 µl
    Yellow
    Log in for availability
    Log in for availability
    Each
    Each - $185.95
    $185.95
    Each
       $185.95
    Supplier Number: FVM04
  • 470344-154
    Micropipette
    Micropipette
    25 µl
    Yellow
    Log in for availability
    Log in for availability
    Each
    Each - $185.95
    $185.95
    Each
       $185.95
    Supplier Number: FVM05
  • 470343-738
    Micropipette
    Micropipette
    50 µl
    Blue
    Log in for availability
    Log in for availability
    Each
    Each - $185.95
    $185.95
    Each
       $185.95
    Supplier Number: FVM06
  • 470344-986
    Micropipette
    Micropipette
    100 µl
    Purple
    Log in for availability
    Log in for availability
    Each
    Each - $185.95
    $185.95
    Each
       $185.95
    Supplier Number: FVM07
  • 470345-892
    Micropipette
    Micropipette
    200 µl
    Purple
    Log in for availability
    Log in for availability
    Each
    Each - $167.05
    $167.05
    Each
       $167.05
    Supplier Number: FVM08
  • 470345-312
    Micropipette
    Micropipette
    250 µl
    Orange
    Log in for availability
    Log in for availability
    Each
    Each - $185.95
    $185.95
    Each
       $185.95
    Supplier Number: FVM09
  • 470344-362
    Micropipette
    Micropipette
    500 µl
    Orange
    Log in for availability
    Log in for availability
    Each
    Each - $182.25
    $182.25
    Each
       $182.25
    Supplier Number: FVM010
  • 470344-426
    Micropipette
    Micropipette
    1000 µl
    Green
    Log in for availability
    Log in for availability
    Each
    Each - $224.05
    $224.05
    Each
       $224.05
    Supplier Number: FVM012
  • 470344-692
    Micropipette
    Micropipette
    2000 µl
    Green
    Log in for availability
    Log in for availability
    Each
    Each - $224.05
    $224.05
    Each
       $224.05
    Supplier Number: FVM013
  • 470345-258
    Micropipette
    Micropipette
    5000 µl
    Green
    Log in for availability
    Log in for availability
    Each
    Each - $224.05
    $224.05
    Each
       $224.05
    Supplier Number: FVM014
  • 470345-196
    Micropipette
    Micropipette
    10000 µl
    Green
    Log in for availability
    Log in for availability
    Each
    Each - $224.05
    $224.05
    Each
       $224.05
    Supplier Number: FVM015
Tips for micropipettes
Product Information
  • Catalog #
    Packed
    Volume
    Availability
    Price
  • 1000 Tips/bag
    0.2 - 10 µl
    Log in for availability
    Log in for availability
    Pack of 1
    Pack of 1 - $127.40
    $127.40
    Pack of 1
       $127.40
Product Information
  • Catalog #
    Packed
    Volume
    Availability
    Price
  • 500 Tips/bag
    200 - 1000 µl
    Log in for availability
    Log in for availability
    Pack of 1
    Pack of 1 - $89.95
    $89.95
    Pack of 1
       $89.95
Product Information
  • Catalog #
    Packed
    Volume
    Availability
    Price
  • 1000 Tips/bag
    2.0 - 200 µl
    Log in for availability
    Log in for availability
    Pack of 1
    Pack of 1 - $89.95
    $89.95
    Pack of 1
       $89.95