PIP® Bouton® Universal Fit PETG Safety Visor
Catalog # 76944-636
Supplier: PIP
Specifications
- Description:Universal fit safety visor
- Material:PETG
- Thickness:0.040"
- Cat. no.:76944-636
Specifications
About this item
Universal Fit PETG (Polyethylene Terephthalate Glycol) Safety Visor with 0.040" thick lens.
- Made with virgin resin for better uniformity of the viewing area
- Universal hole pattern will fit most headgear and hard hat adapters
- Ships with masking on both sides to prevent scratching
PETG (Polyethylene Terephthalate Glycol) safety visor is ideal for liquid splash applications.