ContactUsLinkComponent
Live or Dead™ Yeast Viability Assay Kit
Live or Dead™ Yeast Viability Assay Kit
  76779-024
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
 :  
undefined
Live or Dead™ Yeast Viability Assay Kit
  76779-024
 :  22475
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
 :  
  • Cat. no.:
    76779-024
  • No. of Tests:
    100
Differentiates live yeast with intact membranes will stain fluorescent green, and dead yeast with damaged membranes will stain fluorescent red.
  • robust protocol
  • live yeast will glow green
  • dead yeast will glow red
This kit contains two ready-to-use nucleic acid stains: MycoLight™ Green JJ98, a replacement for SYTO™ 9, and propidium iodide.
Quantitatively distinguishes live and dead yeast within minutes using flow cytometry