ContactUsLinkComponent
Live or Dead™ Yeast CFDA-AM/Propidium Iodide Vitality Kit
Live or Dead™ Yeast CFDA-AM/Propidium Iodide Vitality Kit
  76779-096
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
 :  
undefined
Live or Dead™ Yeast CFDA-AM/Propidium Iodide Vitality Kit
  76779-096
 :  22476
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
 :  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
  • Cat. no.:
    76779-096
  • No. of Tests:
    100
Differentiates live yeast with intact membranes will stain fluorescent green, and dead yeast with damaged membranes will stain fluorescent red.
  • robust protocol
  • live yeast will glow green
  • dead yeast will glow red
The kit uses CFDA-AM and Propidium Iodide. CFDA-AM is a cell permeable esterase substrate.
The acetoxymethyl (AM) group of the substrate will allow CFDA to penetrate cell membranes. Once inside the cells, lipophilic blocking and diacetate groups are cleaved by esterases, resulting in green fluorescence. In contrast, Propidium Iodide only stain the yeasts with damaged membranes resulting in red fluorescence.
Convenient way to test viability of yeasts