Bucculite™ XdU Cell Proliferation Fluorescence Imaging Kit
76779-110
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
:
- Cat. no.:76779-110
- No. of Tests:200
Provides an alternative to anti-BrdU antibody-based assay and EdU click chemistry-based assay
- alternative to anti-BrdU
- extremely sensitive
- deep red fluorescence
The resulted iFluor® 647-labeled DNA formed in cells is imaged with a Cy5 filter set.
Uses XdU which is incorporated into cellular DNA during DNA synthesis