ContactUsLinkComponent
Bucculite™ XdU Cell Proliferation Fluorescence Imaging Kit
Bucculite™ XdU Cell Proliferation Fluorescence Imaging Kit
Catalog # 76779-086
Supplier:  AAT BIOQUEST INC
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  
undefined
Bucculite™ XdU Cell Proliferation Fluorescence Imaging Kit
Catalog # 76779-086
Supplier:  AAT BIOQUEST INC
Supplier Number:  22327
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  
Specifications
  • Cat. no.:
    76779-086
  • No. of Tests:
    200
Specifications
About this item
Provides an alternative to anti-BrdU antibody-based assay and EdU click chemistry-based assay
  • alternative to anti-BrdU
  • extremely sensitive
  • red fluorescence
The resulted iFluor® 555-labeled DNA formed in cells is imaged with a Cy3 filter set.
Uses XdU which is incorporated into cellular DNA during DNA synthesis