NOVAX® Straight Cuff Rubber Insulating Gloves, Red
Supplier: PIP
New Product
Certificates
About this item
Class 0 red rubber insulating gloves with a straight cuff, available in 11 or 14" lengths.
- Contoured shape reduces hand fatigue
- Silicone-free
- Individually numbered for traceability and stamped with factory manufacturing date
Natural rubber construction offers the required dielectric properties, combined with flexibility, strength, and durability.
Certifications: Gloves meet ANSI/ASTM D120 standard and NFPA 70E for use around electrical hazards and arc flash protection.
Specifications
- Catalog No:
- 76944-498
- 76944-500
- 76944-514
- 76944-516
- 76944-510
- 76944-512
- 76944-496
- 76944-508
- 76944-492
- 76944-504
- 76944-494
- 76944-506
- 76944-502
- Ambidextrous/Hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Latex/Latex-free:
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Powdered/Powder-free:
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Material:
- Rubber
- Rubber
- Rubber
- Rubber
- Rubber
- Rubber
- Rubber
- Rubber
- Rubber
- Rubber
- Rubber
- Rubber
- Rubber
- Cuff style:
- Straight
- Straight
- Straight
- Straight
- Straight
- Straight
- Straight
- Straight
- Straight
- Straight
- Straight
- Straight
- Straight
- Sterility:
- Non sterile
- Non sterile
- Non sterile
- Non sterile
- Non sterile
- Non sterile
- Non sterile
- Non sterile
- Non sterile
- Non sterile
- Non sterile
- Non sterile
- Non sterile