To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Description:DNA/chromosome staining lab activity
- Environmentally Preferable:
- Cat. no.:470336-172
- Includes:SDS and instructions
- No. of Uses:5
Specifications
About this item
Prepare stained slides to view plant samples in various stages of development.
- Teacher's manual and study guide are included
Prepare your own squashed stained slide and be able to identify the phases of plant mitosis and chromosomal development. Students will be able to prepare, stain and mount slides using specially prepared onion root tips. Kit contains enough material for 15 groups. Teacher’s Manual and Student Study Guide copymasters are included.