If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Description:Introduction to basic, selective, and differential media
- Cat. no.:470336-152
- Includes:SDS and instructions
- No. of Uses:10
Specifications
About this item
Learn about the different types of media used to grow bacteria.
- Kit contains enough materials for 10 groups
In this activity, students will learn about the different types of media that can be used to grow bacteria. They will predict which media will support the growth of different bacteria, then streak the different media with bacteria cultures to observe their growth. The bacteria growth will be evaluated to determine if students' predictions were correct
10 lyophilized cultures of Escherichia coli; 30 Tryptic soy broth tubes; 40 sterile Petri dishes; 30 sterile pipettors; 40 Cotton swab packs, 2/pack.