VWR® Square Media Biocanisters
Supplier: VWR International
Certificates
About this item
VWR® square containers are ideal for storage of media, laboratory reagents, and biological buffers. Eliminate washing, depyrogenation, and autoclaving steps with sterile and non-pyrogenic containers.
- Made from durable 100% virgin material
- Leakproof bottles and HDPE closures
- Leakproof sealing with integrated lip seal feature
- Sterile and non-pyrogenic, SAL 10⁻⁶
- Low extractables
- Transparency and reduced permeability
All containers are graduated, and feature a molded hand grip for improved ergonomics.
Polycarbonate containers are suitable for use in temperatures ranging from –130 to 121 °C. PET/PETG containers are suitable for use in temperatures ranging from –70 to 60 °C.
Certifications: US FDA 21CRF, USP Class VI compliant resin.
Packaging: Double polybag packed.
Customers Who Bought This Also Bought