Accessories for EXP®2 Stem Trap Kits and Replacement Stems, 2.7 µm HALO®
Catalog # 97056-472
Supplier: Optimize
Specifications
- Description:Stem Trap Holder (Fittings Included)
- Cat. no.:97056-472
- Supplier No.:15-02-03996
Specifications
About this item
The reusable holder are only slightly larger than a standard HPLC fitting. The slim architecture allows it to easily fit into crowded instrument compartments or
to connect directly to tightly-spaced injection ports.