Order Entry
Canada
ContactUsLinkComponent
Anti-AAV8 Rabbit Monoclonal Antibody [clone: 75F2]
Anti-AAV8 Rabbit Monoclonal Antibody [clone: 75F2]
Catalog # 77007-588
Supplier:  Genscript
Anti-AAV8 Rabbit Monoclonal Antibody [clone: 75F2]
Catalog # 77007-588
Supplier:  Genscript
Supplier Number:  A02204-40
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:

  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address

Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Antigen name:
    AAV8
  • Clonality:
    Monoclonal
  • Clone:
    75F2
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Size:
    40 μg
  • Cross adsorption:
    No
  • Form:
    Lyophilized
  • Storage buffer:
    Lyophilized with PBS, pH 7.2, containing 0.02% sodium azide
  • Storage temperature:
    It is stable up to 1 year at −20 °C from date of receipt. The reconstituted antibody can be stored for 2 - 3 weeks at 2...8 °C. For long term storage, aliquot and store at −20 °C or below. Avoid repeated freezing and thawing cycles
  • Concentration:
    Reconstitute the lyophilized powder with deionized water (or equivalent) to a final concentration of 0.5 mg/ml.
  • Immunogen:
    AAV8 Virus-Like Particles (VLPs)
  • Tested applications:
    Dot blot
  • Cat. no.:
    77007-588
  • Packaging:
    Vial

Specifications

About this item

Adeno-associated virus type 8 (AAV8) is a valuable vector for gene therapy. It is one member of the human parvovirus family which depends on co-infection with helper viruses such as adenovirus or herpes virus for efficient reproduction. AAV8 empty capsids are composed of three proteins, VP1, VP2 and VP3 with relative molecular masses of 87, 72 and 62 kDa, respectively.

  • High specificity
  • High affinity
  • High stability

GenScript’s AAV antibody is a useful tool for evaluation of virus expression level, quantification of virus titer, identification of capsid proteins in AAV production, and analysis of AAV immunogenicity.

Type:
Antigen: AAV8
Clonality: Monoclonal
Clone: 75F2
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: