Order Entry
Canada
ContactUsLinkComponent
Anti-Avelumab Mouse Monoclonal Antibody [clone: 12H7]
Anti-Avelumab Mouse Monoclonal Antibody [clone: 12H7]
Catalog # 77007-586
Supplier:  Genscript
Anti-Avelumab Mouse Monoclonal Antibody [clone: 12H7]
Catalog # 77007-586
Supplier:  Genscript
Supplier Number:  A02121-40
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:

  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address

Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Antigen name:
    Avelumab
  • Clonality:
    Monoclonal
  • Clone:
    12H7
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1 kappa
  • Size:
    40 μg
  • Cross adsorption:
    No
  • Form:
    Lyophilized
  • Storage buffer:
    This product is lyophilized with PBS, pH 7.2, containing 0.02% sodium azide
  • Storage temperature:
    The lyophilized product remains stable up to 1 year at −20 °C from date of receipt. Upon reconstitution, it can be stored for 2 - 3 weeks at 2...8 °C or for up to 12 months at −20 °C or below. Avoid repeated freezing and thawing cycles
  • Concentration:
    Reconstitute the lyophilized powder with deionized water (or equivalent) to a final concentration of 0.5 mg/ml.
  • Immunogen:
    Avelumab
  • Cat. no.:
    77007-586
  • Packaging:
    Vial

Specifications

About this item

Avelumab with trade name Bavencio, is an FDA-approved drug to treat Merkel cell carcinoma, urothelial carcinoma, and renal cell carcinoma. Avelumab is a whole monoclonal antibody of isotype IgG1 that binds to the programmed death-ligand 1 (PD-L1) and therefore inhibits binding to its receptor programmed cell death 1 (PD-1). PD-L1 can be highly expressed on certain tumors, which is thought to lead to reduced activation of immune cells (cytotoxic T-cells in particular) that might otherwise recognize and attack the cancer. Inhibition of PD-L1 by Avelumab can remove this inhibitor effect and thereby engender an anti-tumor response. It is one of several ways to block inhibitory signals related to T-cell activation, a more general strategy known as immune checkpoint inhibition. Anti-Avelumab Antibody (12H7), mAb, Mouse is produced from a hybridoma resulting from the fusion of partner and B-lymphocytes obtained from a mouse immunized with Avelumab.

  • High specificity
  • High affinity
  • High stability

GenScript has developed a broad range of off-the-shelf anti-idiotype antibodies against blockbuster drugs to help accelerate your research of biosimilars.

Type:
Antigen: Avelumab
Clonality: Monoclonal
Clone: 12H7
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: