To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
- Antigen name:Etanercept
- Clonality:Monoclonal
- Clone:4H1
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Size:40 μg
- Cross adsorption:No
- Form:Lyophilized
- Storage buffer:This product is lyophilized with PBS, pH 7.2, containing 0.02% sodium azide
- Storage temperature:The lyophilized product remains stable up to 1 year at −20 °C from date of receipt. Upon reconstitution, it can be stored for 2 - 3 weeks at 2...8 °C or for up to 12 months at −20 °C or below. Avoid repeated freezing and thawing cycles
- Concentration:Reconstitute the lyophilized powder with deionized water (or equivalent) to a final concentration of 0.5 mg/ml.
- Immunogen:Etanercept
- Cat. no.:77007-472
- Packaging:Vial
Etanercept with trade name Enbrel, is an FDA-approved drug to treat rheumatoid arthritis, juvenile idiopathic arthritis and psoriatic arthritis, plaque psoriasis and ankylosing spondylitis. Etanercept is a fusion protein made from the combination of two naturally occurring soluble human 75-kilodalton TNF receptors linked to an Fc portion of an IgG1. Etanercept treats autoimmune diseases by interfering with tumor necrosis factor-alpha (TNF alpha, a soluble inflammatory cytokine) by acting as a TNF alpha inhibitor.Anti-Etanercept Antibody (4H1), mAb, Mouse is produced from a hybridoma resulting from the fusion of partner and B-lymphocytes obtained from a mouse immunized with Etanercept.
- High specificity
- High affinity
- High stability
GenScript has developed a broad range of off-the-shelf anti-idiotype antibodies against blockbuster drugs to help accelerate your research of biosimilars.
Type:
Antigen: Etanercept
Clonality: Monoclonal
Clone: 4H1
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: