To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antigen name:Golimumab
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Size:40 μg
- Cross adsorption:No
- Form:Lyophilized
- Storage buffer:This product is lyophilized with PBS, pH 7.2, containing 0.02% sodium azide
- Storage temperature:The lyophilized product remains stable up to 1 year at −20 °C from date of receipt. Upon reconstitution, it can be stored for 2 - 3 weeks at 2...8 °C or for up to 12 months at −20 °C or below. Avoid repeated freezing and thawing cycles
- Concentration:Reconstitute the lyophilized powder with deionized water (or equivalent) to a final concentration of 0.5 mg/ml.
- Immunogen:Golimumab
- Cat. no.:77007-528
- Packaging:Vial
Specifications
About this item
Golimumab with trade name Simponi, is an FDA-approved drug for the treatment of Rheumatoid arthritis (RA), Psoriatic Arthritis (PsA), Ankylosing Spondylitis (AS) and Ulcerative Colitis (UC). It is a human IgG1κ monoclonal antibody that binds to Tumor Necrosis Factor alpha (TNF-alpha), thereby inhibiting the binding to TNF alpha receptors and subsequent generation of inflammatory cytokines. GenScript Anti-Golimumab Antibody, pAb, Rabbit is produced from sera of a rabbit immunized with Golimumab.
- High specificity
- High affinity
- High stability
GenScript has developed a broad range of off-the-shelf anti-idiotype antibodies against blockbuster drugs to help accelerate your research of biosimilars.
Type:
Antigen: Golimumab
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: