About this item
SARS-CoV-2 (Severe acute respiratory syndrome coronavirus 2), also known as 2019-nCoV, is a positive-sense single-stranded RNA virus. It caused coronavirus disease in 2019 (COVID-19). SARS-CoV-2 contains glycosylated spike (S) protein, which is composed of S1 subunit and S2 subunit. The S1 contains a receptor-binding domain (RBD) that can bind to ACE2 receptor on target cells. Lineage B.1.351 (a.k.a. 20H/501Y.V2) is a variant of SARS-CoV-2 (Beta variant). It has multiple mutations in theS1 RBD, including K417N, E484K, N501Y.
- High specificity
- High affinity
- High stability
Genscript offers site-specific neutralizing antibodies, neutralizing antibody standards, and control antibodies.
Type:
Antigen: SARS-CoV-2
Clonality: Monoclonal
Clone: 21F1
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: SARS-CoV-2 beta
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 77007-494
- 77007-602
- Antigen name:
- SARS-CoV-2
- SARS-CoV-2
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- 21F1
- 21F1
- Reactivity:
- SARS-CoV-2 beta
- SARS-CoV-2 beta
- Host:
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- Neutralisation:
- Yes
- Yes