NOVAX® Top Grain Goatskin Leather Protector for Novax® Gloves, Gauntlet Cuff
Supplier: PIP
New Product
Certificates
About this item
Top Grain Goatskin leather construction is the softest, most abrasion-resistant leather and is highly recommended for applications requiring tactile sensitivity.
- Top grain leather is smooth, and provides durability and dexterity
- Split leather gauntlet cuff for added forearm protection and allows for fast removal
Keystone thumb is ergonomic, reduces hand fatigue, and offers superior dexterity and durability. Fourchettes between the fingers enhance the natural contour of the glove for a better fit. Elastic closure sewn inside the glove for safety and comfort. Color-coded hems for easy size identification.
Certifications: Gloves meet ASTM F696-06 (2011) standard.
Specifications
- Catalog No:
- 76944-368
- 76944-370
- 76944-376
- 76944-366
- 76944-374
- 76944-372
- Ambidextrous/Hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Cuff style:
- Gauntlet
- Gauntlet
- Gauntlet
- Gauntlet
- Gauntlet
- Gauntlet
- Sterility:
- Non sterile
- Non sterile
- Non sterile
- Non sterile
- Non sterile
- Non sterile