NOVAX® Electrical Safety Kits, Class 0, Orange Rubber Insulating Gloves
Supplier: PIP
New Product
Certificates
About this item
Electrical safety kits allow users to pick the class that an application requires, the color and size rubber gloves required, and Protective Industrial Products does the rest by assembling a kit that works.
- 1 pair of rubber insulating gloves
- 1 pair of leather protector gloves
- 11" nylon storage bag for gloves
Critical PPE for EV manufacturing. Gloves should meet and/or exceed the requirements of the ASTM D120-14a Standard Specification for Rubber Insulating gloves. Gloves should also be electrically tested following ASTM D120-14a.
Certifications: Gloves should meet and/or exceed the requirements of the ASTM D120-14a.
Specifications
- Catalog No:
- 76944-328
- 76944-334
- 76944-326
- 76944-330
- 76944-340
- 76944-336
- 76944-338
- 76944-332
- Ambidextrous/Hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Sterility:
- Non sterile
- Non sterile
- Non sterile
- Non sterile
- Non sterile
- Non sterile
- Non sterile
- Non sterile
- ASTM No.:
- ASTM D120-14a
- ASTM D120-14a
- ASTM D120-14a
- ASTM D120-14a
- ASTM D120-14a
- ASTM D120-14a
- ASTM D120-14a
- ASTM D120-14a